Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRB3/CD85a/ILT5 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1806N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1806N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1806N100 Supplier R&D Systems Supplier No. FAB1806N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT5/CD85a/LILRB3 Monoclonal specifically detects ILT5/CD85a/LILRB3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ILT5/CD85a/LILRB3
Applications Flow Cytometry
Classification Monoclonal
Clone 222821
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. AAB68668
Gene Alias CD85 antigen-like family member A, CD85a, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3, LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB
Gene Symbols LILRB3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT5/CD85a, Gly24-Glu443, Accession # AAB68668
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 11025
Test Specificity Detects human ILT5/CD85a in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ILT1, rhILT2, rhILT3, rhILT4, rhILT6, rhILT7 or rhILT11 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.