Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRB4/CD85k/ILT3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2425
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2425 100 μg
MAB2425SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2425 Supplier R&D Systems Supplier No. MAB2425
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT3/CD85k/LILRB4 Monoclonal specifically detects ILT3/CD85k/LILRB4 in Human samples. It is validated for Western Blot.

Specifications

Antigen ILT3/CD85k/LILRB4
Applications Western Blot
Classification Monoclonal
Clone 293622
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. ABM86208
Gene Alias CD85 antigen-like family member K, CD85k, CD85k antigen, HM18, ILT3, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5, LIR5LILRB5, LIR-5subfamily B, member 4, member 4
Gene Symbols LILRB4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRB4/CD85k/ILT3 Pro17-His257 Accession # ABM86208
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11006
Test Specificity Detects human LILRB4/CD85k/ILT3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ILT2, 4 5, 6, rhLIR6, or rhLIR8 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.