Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRB4/CD85k/ILT3 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB24251P10
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB24251P10 100 Tests
FAB24251P02 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB24251P10 Supplier R&D Systems Supplier No. FAB24251P100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 7 publications

ILT3/CD85k/LILRB4 Monoclonal specifically detects ILT3/CD85k/LILRB4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ILT3/CD85k/LILRB4
Applications Flow Cytometry
Classification Monoclonal
Clone 293623
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. ABM86208
Gene Alias CD85 antigen-like family member K, CD85k, CD85k antigen, HM18, ILT3, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5, LIR5LILRB5, LIR-5subfamily B, member 4, member 4
Gene Symbols LILRB4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ILT3/CD85k Pro17-His257 Accession # ABM86208
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11006
Test Specificity Detects human ILT3/CD85k in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with with recombinant human (rh) ILT5 is observed. No cross-reactivity with rhILT1, rhILT2, rhILT4, rhILT7, rhILT11, rhLIR-6 or rhLIR-8 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.