Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRB5/CD85c/LIR-8 Antibody, R&D Systems™

Catalog No. MAB30651100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB30651100 100 μg
MAB30651SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB30651100 Supplier R&D Systems Supplier No. MAB30651100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LIR-8/CD85c/LILRB5 Monoclonal antibody specifically detects LIR-8/CD85c/LILRB5 in Human samples. It is validated for Immunocytochemistry

Specifications

Antigen LIR-8/CD85c/LILRB5
Applications Immunocytochemistry
Classification Monoclonal
Clone 1058033
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. O75023
Gene Alias CD85 antigen-like family member C, CD85c, CD85c antigen, Leukocyte immunoglobulin-like receptor 8, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR8, LIR-8, LIR8CD85C, LIR-8leukocyte immunoglobulin-like receptor subfamily B member 5, member 5
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human LILRB5/CD85c/LIR-8, Arg18-His456, Accession # O75023
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10990
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.