Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LIM1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $503.00
Specifications
Antigen | LIM1 |
---|---|
Clone | 320416 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Description
LIM1 Monoclonal specifically detects LIM1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LIM1 | |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
P48742 | |
3975 | |
E. coli-derived recombinant human LIM1 Leu265-Trp406 Accession # P48742 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human LIM1 in direct ELISAs and Western blots. |
320416 | |
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
hLim-1, Homeobox protein Lim-1, LHX1, LIM homeobox 1, LIM homeobox protein 1, LIM-1LIM/homeobox protein Lhx1, LIM1MGC138141, MGC126723 | |
LHX1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title