Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LINGO-1 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB30861A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB30861A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB30861A Supplier R&D Systems Supplier No. FAB30861A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LINGO-1 Monoclonal specifically detects LINGO-1 in Human samples. It is validated for Flow Cytometry.

Order Info

The product is shipped with polar packs.

Specifications

Antigen LINGO-1
Applications Flow Cytometry
Classification Monoclonal
Clone 332237
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_116197
Gene Alias FLJ14594, LERN1, LERN1UNQ201, leucine rich repeat and Ig domain containing 1, leucine rich repeat neuronal 6A, Leucine-rich repeat and immunoglobilin domain-containing protein 1, Leucine-rich repeat neuronal protein 1, Leucine-rich repeat neuronal protein 6A, LINGO1, LRRN6A, LRRN6Aleucine-rich repeat neuronal 6A, MGC17422
Gene Symbols LINGO1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LINGO-1 Thr40-Thr556 Accession # NP_116197
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84894
Test Specificity Detects human LINGO-1 in direct ELISA and Western blot. In direct ELISA and Western blot, approximately 10% cross-reactivity with recombinant human (rh) LINGO-2 is observed and no cross-reactivity with rhLINGO-3 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.