Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lipocalin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1708
This item is not returnable.
View return policy
Description
Lipocalin-1 Monoclonal specifically detects Lipocalin-1 in Human samples. It is validated for Western Blot.Specifications
Lipocalin-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P31025 | |
LCN1 | |
Mouse myeloma cell line NS0-derived recombinant human Lipocalin-1 His19-Asp176 Accession # P31025 | |
500 μg | |
Primary | |
Detects human Lipocalin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human Lipocalin-2. | |
Human | |
Purified |
Western Blot | |
223908 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
LCN1, lipocalin 1 (tear prealbumin), lipocalin 1-like 2, lipocalin-1, Lipocalin1, MGC71975, PMFA, Tear lipocalin, Tear prealbumin, Tlc, TPVon Ebner gland protein, VEG protein, VEGP, VEGPlipocalin 1 (protein migrating faster than albumin, tear prealbumin) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3933 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction