Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Lipocalin-1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB170575 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170575 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170575 Supplier Novus Biologicals™ Supplier No. NBP318729
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 1.25 - 80 ng/mL (example only; lot dependent)
Assay Sensitivity 0.65 ng/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture, Plasma, Saliva, Serum, Urine
For Use With (Application) ELISA
Gene ID (Entrez) 3933
Gene Symbol LCN1
Quantity 1 Kit
Research Discipline Cancer
Storage Requirements Storage is content dependent.
Synonym lipocalin 1 (tear prealbumin), lipocalin 1-like 2, lipocalin-1, MGC71975, PMFA, Tear lipocalin, Tear prealbumin, Tlc, TPVon Ebner gland protein, VEG protein, VEGPlipocalin 1 (protein migrating faster than albumin, tear prealbumin)
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.