Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lipocalin-2/NGAL Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
$214.00 - $2020.00
Specifications
Antigen | Lipocalin-2/NGAL |
---|---|
Clone | 220310 |
Concentration | LYOPH |
Dilution | Western Blot 0.2-1 ug/mL, Immunohistochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1757SP
![]() |
R&D Systems
MAB1757SP |
25 μg |
Each for $214.00
|
|
|||||
MAB1757100
![]() |
R&D Systems
MAB1757100 |
100 μg |
Each for $549.50
|
|
|||||
MAB1757500
![]() |
R&D Systems
MAB1757500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Lipocalin-2/NGAL | |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin | |
LCN2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
220310 | |
Western Blot 0.2-1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Rat | |
Human | |
P80188 | |
3934 | |
Mouse myeloma cell line NS0-derived recombinant human Lipocalin-2/NGAL Gln21-Gly198 Accession # P80188 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Lipocalin-2/NGAL in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) Lipocalin-1 or rmLipocalin-2. This antibody also recognizes human Lipocalin-2/MMP-9 complexes in Western blots under non-reducing conditions. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title