Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LITAF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4695SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4695SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4695SP Supplier R&D Systems Supplier No. AF4695SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

LITAF Polyclonal specifically detects LITAF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen LITAF
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99732
Gene Alias CMT1C, FLJ38636, lipopolysaccharide-induced TNF factor, lipopolysaccharide-induced TNF-alpha factor, lipopolysaccharide-induced tumor necrosis factor-alpha factor, LPS-induced TNF-alpha factor, MGC116698, MGC116700, MGC125275, MGC125276, p53-induced gene 7 protein, PIG7, PIG7MGC116701, SIMPLE, SIMPLEMGC125274, Small integral membrane protein of lysosome/late endosome, TP53I7, tumor protein p53 inducible protein 7
Gene Symbols LITAF
Host Species Goat
Immunogen E. coli-derived recombinant human LITAF Met1-Ala111 Accession # Q99732
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9516
Test Specificity Detects human LITAF in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.