Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LITAF Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4695
This item is not returnable.
View return policy
Description
LITAF Polyclonal specifically detects LITAF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
LITAF | |
Polyclonal | |
Unconjugated | |
Q99732 | |
LITAF | |
E. coli-derived recombinant human LITAF Met1-Ala111 Accession # Q99732 | |
100 μg | |
Primary | |
Detects human LITAF in Western blots. | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
CMT1C, FLJ38636, lipopolysaccharide-induced TNF factor, lipopolysaccharide-induced TNF-alpha factor, lipopolysaccharide-induced tumor necrosis factor-alpha factor, LPS-induced TNF-alpha factor, MGC116698, MGC116700, MGC125275, MGC125276, p53-induced gene 7 protein, PIG7, PIG7MGC116701, SIMPLE, SIMPLEMGC125274, Small integral membrane protein of lysosome/late endosome, TP53I7, tumor protein p53 inducible protein 7 | |
Goat | |
Affinity Purified | |
RUO | |
9516 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction