Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Liver Antigen Monoclonal Antibody (EP-1)
GREENER_CHOICE

Catalog No. PIMA533330
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533330 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533330 Supplier Invitrogen™ Supplier No. MA533330
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monoclonal antibody EP-1 stains human liver canaliculi and a subset of hepatocellular carcinomas.

Human liver antigen is a gene target that plays a crucial role in liver function and metabolism. The liver is a vital organ responsible for numerous functions, including detoxification, synthesis of proteins, and bile production. Liver antigens are proteins expressed specifically in liver cells and are involved in maintaining liver health and homeostasis. Studying liver antigens can provide valuable insights into liver diseases, such as hepatitis, cirrhosis, and liver cancer. Understanding the genetic variations and expression patterns of liver antigens can aid in the development of diagnostic tests, therapeutic interventions, and personalized treatment strategies for liver-related conditions. Furthermore, investigating liver antigens can contribute to advancements in liver transplantation and regenerative medicine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Liver Antigen
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone EP-1
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ELAVL4
Gene Accession No. P26378
Gene Alias Elav; ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV like neuron-specific RNA binding protein 4; ELAVL4; ELAV-like protein 4; Hu antigen D; HU-antigen D; Hud; Paraneoplastic encephalomyelitis antigen HuD; PNEM; RGD1561943; r-HuD; RNA-binding protein HUD3
Gene Symbols ELAVL4
Host Species Mouse
Immunogen SK-H1A9-2 human hepatocarcinoma cells.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1996
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.