Learn More
R&D Systems™ Human Liver Whole Tissue Lysate (Adult Whole Normal)
Shop All Novus Biologicals Products

Specifications
Specifications
| Preparation Method | Western Blot Electrophoresis |
| Quantity | 1 mg |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| Product Line | Novus |
| Target | Liver |
| Synonym | Liver, Liver lysate, liver lysates |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.