Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Liver Whole Tissue Lysate (Adult Whole Primary and Metastatic Tumor Paired)
SDP

Catalog No. NBL126530 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBL126530 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBL126530 Supplier Novus Biologicals™ Supplier No. NBL126530

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

The Human Liver Whole Tissue Lysate (Adult Whole Primary and Metastatic Tumor Paired) is derived from human.

Specifications

Tissue Digestive/Waste
Quantity 1 Set
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Matched Pair - Total Protein - Human Primary Tumor and Metastatic Tumor Tissue (PM): Liver
Cellular Fraction Whole
Tissue Condition Primary and Metastatic Tumor Paired
Cross Reactivity Human
Host Species Human
Product Type Human, Liver Whole Tissue Lysate (Adult Primary and Metastatic Tumor Paired)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.