Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Liver Whole Tissue Lysate (Adult Whole Primary Tumor and Normal Paired)
SDP

Catalog No. NB82089076 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB82089076 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB82089076 Supplier R&D Systems™ Supplier No. NB82089076
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Applications: Western Blot, Immunoprecipitation

The Human Liver Whole Tissue Lysate (Adult Whole Primary Tumor and Normal Paired) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.

Specifications

Quantity 1 Set
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Test Specificity Matched Pair - Total Protein - Human Primary Tumor and Normal Tissue (PP): Liver
Cellular Fraction Whole
Tissue Condition Primary Tumor and Normal Paired
Cross Reactivity Human
Host Species Human
Concentration 5 mg/mL
Product Type Human, Liver Whole Tissue Lysate (Adult Primary Tumor and Normal Paired)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Product Line Novus
Protein State Native
For Use With (Application) Western Blot
Tissue Digestive/Waste
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.