Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LMP2/PSMB9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7709
This item is not returnable.
View return policy
Description
LMP2/PSMB9 Monoclonal specifically detects LMP2/PSMB9 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
LMP2/PSMB9 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P28065 | |
PSMB9 | |
E. coli-derived recombinant human LMP2/PSMB9 Thr21-Glu219 Accession # P28065 | |
100 μg | |
Primary | |
Detects human LMP2/PSMB9 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human PSMB6 is observed. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
792520 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta1i, LMP2MGC70470, Low molecular mass protein 2, Macropain chain 7, Multicatalytic endopeptidase complex chain 7, proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalpeptidase 2), proteasome catalytic subunit 1i, Proteasome chain 7, proteasome subunit beta 6i, proteasome subunit beta type-9, Proteasome subunit beta-1i, proteasome-related gene 2, PSMB6iproteasome (prosome, macropain) subunit, beta type, 9 (large multifunctionalprotease 2), PSMB9, Really interesting new gene 12 protein, RING12, RING12EC 3.4.25.1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5698 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction