Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LMP7/PSMB8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7710
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7710 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7710 Supplier R&D Systems Supplier No. MAB7710
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LMP7/PSMB8 Monoclonal specifically detects LMP7/PSMB8 in Human samples. It is validated for Western Blot.

Specifications

Antigen LMP7/PSMB8
Applications Western Blot
Classification Monoclonal
Clone 832913
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P28062
Gene Alias ALDD, beta5i, D6S216E, EC 3.4.25.1, JMP, LMP7MGC1491, Low molecular mass protein 7, low molecular weight protein 7, Macropain subunit C13, Multicatalytic endopeptidase complex subunit C13, NKJO, protease component C13, proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctionalpeptidase 7), proteasome catalytic subunit 3i, Proteasome component C13, proteasome subunit beta 5i, proteasome subunit beta type-8, Proteasome subunit beta-5i, proteasome subunit Y2, proteasome-related gene 7, PSMB5i, PSMB5iD6S216, PSMB8, Really interesting new gene 10 protein, RING10, RING10proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctionalprotease 7), Y2
Gene Symbols PSMB8
Host Species Mouse
Immunogen E. coli-derived recombinant human LMP7/PSMB8 Thr73-Gln276 Accession # P28062
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5696
Test Specificity Detects human LMP7/PSMB8 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human LMPX/PSMB5 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.