Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LMTK2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5147SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5147SP 25 μg
MAB5147 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5147SP Supplier R&D Systems Supplier No. MAB5147SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

LMTK2 Monoclonal specifically detects LMTK2 in Human samples. It is validated for Western Blot.

Specifications

Antigen LMTK2
Applications Western Blot
Classification Monoclonal
Clone 465222
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8IWU2
Gene Alias AATK2, AATYK2FLJ46659, Apoptosis-associated tyrosine kinase 2, Brain-enriched kinase, BREK, BREKCDK5/p35-regulated kinase, cprk, EC 2.7.11, KIAA1079hBREK, kinase phosphatase inhibitor 2, Kinase/phosphatase/inhibitor 2, KPI-2, KPI2EC 2.7.11.1, lemur tyrosine kinase 2Serine/threonine-protein kinase KPI-2, LMR2, LMR2cyclin-dependent kinase 5/p35-regulated kinase, serine/threonine-protein kinase LMTK2
Gene Symbols LMTK2
Host Species Rat
Immunogen E. coli-derived recombinant human LMTK2 Ala93-Tyr284 Accession # Q8IWU2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22853
Test Specificity Detects human LMTK2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human AATK is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.