Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LOX-1/OLR1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1798
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1798 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1798 Supplier R&D Systems Supplier No. AF1798
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

LOX-1/OLR1 Polyclonal specifically detects LOX-1/OLR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.

Specifications

Antigen LOX-1/OLR1
Applications Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction
Gene Accession No. P78380
Gene Alias CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1
Gene Symbols OLR1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human LOX-1 (R&D Systems, Catalog # 1798-LX) Ser61-Gln273 Accession # P78380
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4973
Test Specificity Selected for its ability to block receptor-ligand interaction in a functional ELISA assay. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse LOX-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.