Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LPAR2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6396
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6396 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6396 Supplier R&D Systems Supplier No. AF6396
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

LPAR2/LPA2/EDG-4 Polyclonal specifically detects LPAR2/LPA2/EDG-4 in Human samples. It is validated for Western Blot.

Specifications

Antigen LPAR2/LPA2/EDG-4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q9HBW0
Gene Alias EDG4, EDG-4, EDG4FLJ93869, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor4,LPA receptor EDG4, G protein-coupled receptor, LPA2, LPA-2, LPA2LPA receptor 2, LPAR2, lysophosphatidic acid receptor 2, lysophosphatidic acid receptor EDG4, Lysophosphatidic acid receptor Edg-4
Gene Symbols LPAR2
Host Species Sheep
Immunogen E. coli-derived recombinant human LPAR2 Met1-Val33, His91-Gly103, Ser168-Ser188, Asp264-Tyr279 Accession # Q9HBW0
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9170
Test Specificity Detects human LPAR2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human LPAR1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.