Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LPAR4/LPA4 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10217P100
This item is not returnable.
View return policy
Description
P2Y9 Monoclonal specifically detects P2Y9 in Human samples. It is validated for Flow Cytometry.Specifications
P2Y9 | |
Monoclonal | |
PE | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
G protein-coupled receptor 23, GPCR23, GPR23, GPR23P2Y purinoceptor 9, LPA4LPA-4, lysophosphatidic acid receptor 4, P2RY9, P2RY9G-protein coupled receptor 23, P2Y5-LIKE, P2Y5-like receptor, P2Y9, P2Y9LPA receptor 4, Purinergic receptor 9 | |
Human embryonic kidney cell line HEK293-derived transfected with human LPAR4/LPA4, Accession # Q99677 | |
100 Tests | |
Primary | |
Detects human LPAR4/LPA4 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Flow Cytometry | |
561203 | |
Flow Cytometry 10 uL/10^6 cells | |
Q99677 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2846 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction