Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LPAR5/LPA5/GPR92 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB10297N100UG
This item is not returnable.
View return policy
Description
LPAR5/LPA5/GPR92 Monoclonal specifically detects LPAR5/LPA5/GPR92 in Human samples. It is validated for Flow Cytometry.Specifications
LPAR5/LPA5/GPR92 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
G protein-coupled receptor 92, GPR92, GPR92G-protein coupled receptor 92, GPR93, GPR93G-protein coupled receptor 93, KPG_010, LPA receptor 5, LPA5, LPA5LPA-5, lysophosphatidic acid receptor 5 | |
NS0 mouse myeloma cell line transfected with human LPAR5/LPA5/GPR92, Accession # Q9H1C0 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
459812 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9H1C0 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
57121 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction