Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LRH-1/NR5A2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. PPH232500
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPH232500 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPH232500 Supplier R&D Systems Supplier No. PPH232500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 8 publications

LRH-1/NR5A2 Monoclonal specifically detects LRH-1/NR5A2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Direct ELISA.

Specifications

Antigen LRH-1/NR5A2
Applications Western Blot, Immunoprecipitation, ELISA
Classification Monoclonal
Clone H2325
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation, Direct ELISA
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
Gene Alias Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1 beta, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH1, LRH-1CPF, NR5A2, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2
Gene Symbols NR5A2
Host Species Mouse
Immunogen Recombinant human LRH-1/NR5A2 aa 161-280
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2494
Test Specificity This antibody specifically recognizes human LRH-1 alpha and LRH-1 beta and cross-reacts with mouse and rat LRH-1. Not yet tested in other species.
Target Species Human
Content And Storage This antibody is stable for greater than six months when held at -20 degreesC in a manual defrost freezer or at -70 degreesC. Upon thawing, the antibody can be stored at 2-8 degreesC for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.