Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LRIG1 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB7498V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7498V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7498V100 Supplier R&D Systems Supplier No. FAB7498V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRIG1 Monoclonal specifically detects LRIG1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LRIG1
Applications Flow Cytometry
Classification Monoclonal
Clone 789211
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q96JA1
Gene Alias D6Bwg0781e, Img, LIG1, LIG-1
Gene Symbols LRIG1
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human LRIG1 Ala35-Ser779 Accession # Q96JA1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26018
Test Specificity Detects human LRIG1 in ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse LRIG1 and recombinant human LRIG3 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.