Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LRIG1 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8140G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8140G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8140G Supplier R&D Systems Supplier No. FAB8140G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

LRIG1 Polyclonal specifically detects LRIG1 in Human samples. It is validated for Flow Cytometry.

Order Info

The product is shipped with polar packs.

Specifications

Antigen LRIG1
Applications Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q96JA1
Gene Alias D6Bwg0781e, Img, LIG1, LIG-1
Gene Symbols LRIG1
Host Species Sheep
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human LRIG1 Ala35-Ser779 Accession # Q96JA1
Purification Method Antigen Affinity-purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26018
Test Specificity Detects human LRIG1 in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse LRIG1 and and less 1% cross-reactivity with recombinant human LRIG3 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.