Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LRIG1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7498SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7498SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7498SP Supplier R&D Systems Supplier No. AF7498SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

LRIG1 Polyclonal specifically detects LRIG1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen LRIG1
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96JA1
Gene Alias D6Bwg0781e, Img, LIG1, LIG-1
Gene Symbols LRIG1
Host Species Sheep
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human LRIG1 Ala35-Ser779 Accession # Q96JA1
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26018
Test Specificity Detects human LRIG1 in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse LRIG1 and and less than 1% cross-reactivity with recombinant human LRIG3 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.