Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LRIG1 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7498A
missing translation for 'nonReturnableMessage'
missing translation for 'returnPolicyLabel'
Description
LRIG1 Monoclonal specifically detects LRIG1 in Human samples. It is validated for Flow Cytometry.Order Info
The product is shipped with polar packs.
Specifications
LRIG1 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
D6Bwg0781e, Img, LIG1, LIG-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
26018 | |
Human | |
Purified |
Flow Cytometry | |
789211 | |
Flow Cytometry 10 uL/10^6 cells | |
Q96JA1 | |
LRIG1 | |
HEK293 human embryonic kidney cell line transfected with human LRIG1 Ala35-Ser779 Accession # Q96JA1 | |
100 Tests | |
Primary | |
Detects human LRIG1 in ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse LRIG1 and recombinant human LRIG3 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction