Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LRIG2 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1941V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1941V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1941V100 Supplier R&D Systems Supplier No. FAB1941V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRIG2 Monoclonal specifically detects LRIG2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LRIG2
Applications Flow Cytometry
Classification Monoclonal
Clone 990310
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 μg/10^6 cells
Formulation Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O94898
Gene Alias KIAA0806, leucine-rich repeats and immunoglobulin-like domains 2, leucine-rich repeats and immunoglobulin-like domains protein 2, LIG2, UFS2
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human LRIG-2 Gly41-Thr805 Accession # O94898
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 9860
Test Specificity Detects human LRIG-2 in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.