Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LRIG3 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3495V100UG
This item is not returnable.
View return policy
Description
LRIG3 Monoclonal specifically detects LRIG3 in Human samples. It is validated for Flow Cytometry.Specifications
LRIG3 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q6UXM1 | |
LRIG3 | |
Mouse myeloma cell line NS0-derived recombinant human LRIG3 Asp28-Thr807 Accession # Q6UXM1 | |
RUO | |
121227 | |
Human | |
IgG2b |
Flow Cytometry | |
814834 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
FLJ90440, KIAA3016, leucine-rich repeats and immunoglobulin-like domains 3, leucine-rich repeats and immunoglobulin-like domains protein 3, LIG-3, LIG3FLJ26573, LIRG3 | |
Mouse | |
100 μg | |
Primary | |
Detects human LRIG3 in direct ELISAs and Western blots. Detects mouse LRIG3 in Western blots. Mouse LRIG3 detection in Flow Cytometry was not tested. In direct ELISAs, no cross-reactivity with recombinant human LRIG1 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction