Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LRIG3 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3495T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3495T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3495T100 Supplier R&D Systems Supplier No. FAB3495T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRIG3 Monoclonal specifically detects LRIG3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LRIG3
Applications Flow Cytometry
Classification Monoclonal
Clone 814834
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q6UXM1
Gene Alias FLJ90440, KIAA3016, leucine-rich repeats and immunoglobulin-like domains 3, leucine-rich repeats and immunoglobulin-like domains protein 3, LIG-3, LIG3FLJ26573, LIRG3
Gene Symbols LRIG3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LRIG3 Asp28-Thr807 Accession # Q6UXM1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121227
Test Specificity Detects human LRIG3 in direct ELISAs and Western blots. Detects mouse LRIG3 in Western blots. Mouse LRIG3 detection in Flow Cytometry was not tested. In direct ELISAs, no cross-reactivity with recombinant human LRIG1 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.