Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LRIG3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3495SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3495SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3495SP Supplier R&D Systems Supplier No. MAB3495SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRIG3 Monoclonal specifically detects LRIG3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen LRIG3
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 814834
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6UXM1
Gene Alias FLJ90440, KIAA3016, leucine-rich repeats and immunoglobulin-like domains 3, leucine-rich repeats and immunoglobulin-like domains protein 3, LIG-3, LIG3FLJ26573, LIRG3
Gene Symbols LRIG3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LRIG3 Asp28-Thr807 Accession # Q6UXM1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121227
Test Specificity Detects human LRIG3 in direct ELISAs and Western blots. Detects mouse LRIG3 in Western blots. Mouse LRIG3 detection in Flow Cytometry was not tested. In direct ELISAs, no cross-reactivity with recombinant human LRIG1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.