Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LRMP Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6869SP
This item is not returnable.
View return policy
Description
LRMP/Jaw1 Polyclonal specifically detects LRMP/Jaw1 in Human samples. It is validated for Western Blot.Specifications
LRMP/Jaw1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Jaw1, JAW1Protein Jaw1, lymphoid-restricted membrane protein | |
Sheep | |
Affinity Purified | |
RUO | |
4033 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
AAI26418 | |
LRMP | |
E. coli-derived recombinant human LRMP Gln143-Pro292 Accession # AAI26418 | |
25 μg | |
Primary | |
Detects human LRMP in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction