Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LRRC32/GARP Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB6055R100UG
This item is not returnable.
View return policy
Description
LRRC32/GARP Monoclonal specifically detects LRRC32/GARP in Human samples. It is validated for Flow Cytometry.Specifications
LRRC32/GARP | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q14392 | |
LRRC32 | |
recombinant human LRRC32/GARP Accession # Q14392 | |
RUO | |
2615 | |
Human | |
IgG2a |
Flow Cytometry | |
855151 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
D11S833E, D11S833Eleucine-rich repeat-containing protein 32, GARP, GARPGarpin, Garpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
Rat | |
100 μg | |
Primary | |
Detects human LRRC32/GARP in ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse LRRC32 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction