Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LRRC32/GARP Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6055S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6055S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6055S100 Supplier R&D Systems Supplier No. FAB6055S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

LRRC32/GARP Monoclonal specifically detects LRRC32/GARP in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LRRC32/GARP
Applications Flow Cytometry
Classification Monoclonal
Clone 855151
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q14392
Gene Alias D11S833E, D11S833Eleucine-rich repeat-containing protein 32, GARP, GARPGarpin, Garpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32
Gene Symbols LRRC32
Host Species Rat
Immunogen recombinant human LRRC32/GARP Accession # Q14392
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2615
Test Specificity Detects human LRRC32/GARP in ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse LRRC32 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.