Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LRRC3B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7635SP
This item is not returnable.
View return policy
Description
LRRC3B/LRP15 Monoclonal specifically detects LRRC3B/LRP15 in Human samples. It is validated for Western Blot.Specifications
LRRC3B/LRP15 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
Q96PB8 | |
LRRC3B | |
Chinese hamster ovary cell line CHO-derived recombinant human LRRC3B Phe28-Ala205 Accession # Q96PB8 | |
25 μg | |
Primary | |
Detects human LRRC3B in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
758315 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
leucine rich repeat containing 3B, leucine-rich repeat-containing protein 3B, LRP15, LRP15Leucine-rich repeat protein LRP15, MGC102927 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
116135 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction