Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LRRC3B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7635
This item is not returnable.
View return policy
Description
LRRC3B/LRP15 Monoclonal specifically detects LRRC3B/LRP15 in Human samples. It is validated for Western Blot.Specifications
LRRC3B/LRP15 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
Q96PB8 | |
LRRC3B | |
Chinese hamster ovary cell line CHO-derived recombinant human LRRC3B Phe28-Ala205 Accession # Q96PB8 | |
100 μg | |
Primary | |
Detects human LRRC3B in direct ELISAs and Western blots. | |
Human | |
Purified |
Western Blot | |
758315 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
leucine rich repeat containing 3B, leucine-rich repeat-containing protein 3B, LRP15, LRP15Leucine-rich repeat protein LRP15, MGC102927 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
116135 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction