Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LRRC4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4995
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4995 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4995 Supplier R&D Systems Supplier No. AF4995
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

LRRC4 Polyclonal specifically detects LRRC4 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen LRRC4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q9HBW1
Gene Alias BAG, Brain Tumor Associated Protein LRRC4, Brain Tumor-Associated Protein BAG, Leucine Rich Repeat Containing 4, Leucine-Rich Repeat-Containing 4, Leucine-Rich Repeat-Containing Protein 4, NAG14, Nasopharyngeal Carcinoma-Associated Gene 14 Protein, Netrin-G2 Ligand, NGL-2
Gene Symbols LRRC4
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human LRRC4 Ala39-Lys527 Accession # Q9HBW1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64101
Test Specificity Detects human LRRC4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human LRRC3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.