Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LRRTM1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4897
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4897 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4897 Supplier R&D Systems Supplier No. AF4897
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

LRRTM1 Polyclonal specifically detects LRRTM1 in Human samples. It is validated for Western Blot.

Specifications

Antigen LRRTM1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q86UE6
Gene Alias FLJ32082, leucine rich repeat transmembrane neuronal 1, leucine-rich repeat transmembrane neuronal 1 protein, leucine-rich repeat transmembrane neuronal protein 1
Gene Symbols LRRTM1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human LRRTM1 Ala35-Ile425 Accession # Q86UE6
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347730
Test Specificity Detects human LRRTM1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross reactivity with recombinant human (rh) LRRTM2 is observed, and less than 5% cross reactivity rhLRRTM3 and rhLRRTM4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.