Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LRTM1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB10046R100UG
This item is not returnable.
View return policy
Description
LRTM1 Monoclonal specifically detects LRTM1 in Human samples. It is validated for Flow Cytometry.Specifications
LRTM1 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
HT017, leucine-rich repeat and transmembrane domain-containing protein 1, leucine-rich repeats and transmembrane domains 1 | |
Synthetic peptide containing human LRTM1, Accession No. Q9HBL6 | |
100 μg | |
57408.0 | |
Human | |
Supernatant |
Flow Cytometry | |
2381D | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9HBL6 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human LRTM1 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction