Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LSECtin/CLEC4G Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2947U100UG
This item is not returnable.
View return policy
Description
LSECtin/CLEC4G Monoclonal specifically detects LSECtin/CLEC4G in Human samples. It is validated for Flow Cytometry.Specifications
LSECtin/CLEC4G | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CLEC4G, C-type lectin domain family 4 member G, C-type lectin domain family 4, member G, C-type lectin superfamily 4, member G, DTTR431, liver and lymph node sinusoidal endothelial cell C-type lectin, LP2698, LSECtin, Q6UXB4, UNQ431 | |
NS0 mouse myeloma cell line transfected with LSECtin/CLEC4G Ser54-Cys293 Accession # Q6UXB4 | |
RUO | |
339390 | |
Human | |
IgG2a |
Flow Cytometry | |
845404 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q6UXB4 | |
Mouse | |
100 μg | |
Primary | |
Detects human LSECtin/CLEC4G in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction