Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lumican Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Supplier: R&D Systems AF2846
This item is not returnable.
View return policy
Description
Lumican Polyclonal specifically detects Lumican in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Lumican | |
Polyclonal | |
Unconjugated | |
P51884 | |
LUM | |
Mouse myeloma cell line NS0-derived recombinant human Lumican Gln19-Asn338 Accession # P51884 | |
100 μg | |
Primary | |
Detects human Lumican in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse Lumican is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
KSPG lumican, LDC, LUM, lumican, lumican proteoglycan, SLRR2D, SLRR2DLDCKeratan sulfate proteoglycan lumican | |
Goat | |
Affinity Purified | |
RUO | |
4060 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction