Learn More
Novus Biologicals™ Human Lung Whole Tissue Lysate (Adult Whole Primary and Metastatic Tumor Paired)
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Quantity | 1 Set |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Matched Pair - Total Protein - Human Primary Tumor and Metastatic Tumor Tissue (PM): Lung |
| Cellular Fraction | Whole |
| Tissue Condition | Primary and Metastatic Tumor Paired |
| Cross Reactivity | Human |
| Host Species | Human |
| Product Type | Human Lung Whole Tissue Lysate (Adult Primary and Metastatic Tumor Paired) |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Life Stage | Adult |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.