Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LXR alpha/NR1H3 Antibody (Clone K8607), R&D Systems™
GREENER_CHOICE

Catalog No. PPK860700
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPK860700 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPK860700 Supplier R&D Systems Supplier No. PPK860700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

LXR alpha/NR1H3 Monoclonal specifically detects LXR alpha/NR1H3 in Human samples. It is validated for Western Blot, Immunoprecipitation, Gel Supershift Assay.

Specifications

Antigen LXR alpha/NR1H3
Applications Western Blot, Immunoprecipitation, Gel Shift
Classification Monoclonal
Clone K8607
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation, Gel Supershift Assay
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
Gene Alias alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, NR1H3, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1
Gene Symbols NR1H3
Host Species Mouse
Immunogen Recombinant human LXRGlc alpha (amino acids 2 - 97) and mouse myeloma cells (NS-1)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10062
Target Species Human
Content And Storage Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.