Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Lymphocytic Marker Monoclonal Antibody (IPO-74)
GREENER_CHOICE

Catalog No. PIMA533315
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533315 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533315 Supplier Invitrogen™ Supplier No. MA533315
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IPO-74 reacts with human leukemia cell line HL-60 and shows a band at MW 127 kDa (reduced) both in immunoprecipitation and Western blotting. In peripheral blood only subsets of B cells, T cells, neutrophilic and eosinophilic granulocytes are positive. All monocytes are positive. Cell lines U937 and K562 are positive, while Daudy, Raji, Jurkat and Molt-4 are negative, as are IL2-dependent T cell lines/clones.

The lymphocytic marker is a gene target that plays a crucial role in the identification and characterization of lymphocytes, a type of white blood cell. Lymphocytes are key components of the immune system and are responsible for defending the body against infections and diseases. The lymphocytic marker gene helps in distinguishing lymphocytes from other cell types and enables researchers to study their function and behavior. By targeting this gene, scientists can gain insights into the immune response, develop diagnostic tools, and design therapeutic strategies for various immune-related disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Lymphocytic Marker
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone IPO-74
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ELAVL4
Gene Accession No. P26378
Gene Alias Elav; ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV like neuron-specific RNA binding protein 4; ELAVL4; ELAV-like protein 4; Hu antigen D; HU-antigen D; Hud; Paraneoplastic encephalomyelitis antigen HuD; PNEM; RGD1561943; r-HuD; RNA-binding protein HUD3
Gene Symbols ELAVL4
Host Species Mouse
Immunogen Human leukemia cell line HL-60.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1996
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.