Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lymphotoxin-alpha/TNF-beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB211
This item is not returnable.
View return policy
Description
Lymphotoxin-alpha/TNF-beta Monoclonal specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Neutralization, Immunocytochemistry.Specifications
Lymphotoxin-alpha/TNF-beta | |
Monoclonal | |
LYOPH | |
Neutralization 0.1-0.2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P01374 | |
LTA | |
E. coli-derived recombinant human Lymphotoxin-alpha/TNF-beta Leu35-Leu205 Accession # P01374 | |
500 μg | |
Primary | |
Detects human Lymphotoxin-alpha/TNF-beta in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TNF-alpha is observed. Neutralizes the biological activity of recombinant human TNF-beta . Will not neutralize the biological activity of recombinant human TNF-alpha or recombinant mouse TNF-alpha . | |
Human | |
Purified |
Neutralization, Immunocytochemistry | |
5802 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
LT, LTA, LT-alpha, Lymphotoxin alpha, lymphotoxin alpha (TNF superfamily, member 1), Lymphotoxinalpha, tnfb, TNF-beta, TNFBlymphotoxin-alpha, TNFSF1, TNFSF1B, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
4049 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction