Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lymphotoxin-alpha/TNF-beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $2070.00
Specifications
| Antigen | Lymphotoxin-alpha/TNF-beta |
|---|---|
| Clone | 5807R |
| Dilution | Neutralization 8-48 ng/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB621RSP
![]() |
R&D Systems
MAB621RSP |
25 μg |
Each for $214.00
|
|
|||||
MAB621R100
![]() |
R&D Systems
MAB621R100 |
100 μg |
Each for $561.50
|
|
|||||
MAB621R500
![]() |
R&D Systems
MAB621R500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
Lymphotoxin-alpha/TNF-beta Monoclonal specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| Lymphotoxin-alpha/TNF-beta | |
| Neutralization 8-48 ng/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| LT, LTA, LT-alpha, Lymphotoxin alpha, lymphotoxin alpha (TNF superfamily, member 1), Lymphotoxinalpha, tnfb, TNF-beta, TNFBlymphotoxin-alpha, TNFSF1, TNFSF1B, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 | |
| E. coli-derived recombinant human Lymphotoxin-alpha/TNF-beta | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Lymphotoxin-alpha/TNF-beta in ELISAs. |
| 5807R | |
| Neutralization, ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 4049 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title