Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lymphotoxin-alpha/TNF-beta Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AB211NA
This item is not returnable.
View return policy
Description
Lymphotoxin-alpha/TNF-beta Polyclonal specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Western Blot, Neutralization.Specifications
Lymphotoxin-alpha/TNF-beta | |
Polyclonal | |
Unconjugated | |
P01374 | |
LTA | |
E. coli-derived recombinant human Lymphotoxin-alpha/TNF-beta Leu35-Leu205 Accession # P01374 | |
1 mg | |
Primary | |
Detects human Lymphotoxin-alpha/TNF-beta in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human TNF-alpha and recombinant mouse TNF-alpha is observed. Neutralizes the biological activity of recombinant human (rh) TNF-beta and natural human TNF-beta , but will not neutralize the biological activity of rhTNF-alpha or recombinant mouse TNF-alpha . | |
Human | |
Purified |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 1 ug/mL, Neutralization 0.2-0.4 ug/mL | |
LT, LTA, LT-alpha, Lymphotoxin alpha, lymphotoxin alpha (TNF superfamily, member 1), Lymphotoxinalpha, tnfb, TNF-beta, TNFBlymphotoxin-alpha, TNFSF1, TNFSF1B, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 | |
Goat | |
Protein A or G purified | |
RUO | |
4049 | |
Reconstitute at 1 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction