Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Lymphotoxin-alpha/TNF-beta ELISA Kit (Colorimetric)
SDP

Catalog No. NBP183730 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP183730 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP183730 Supplier Novus Biologicals™ Supplier No. NBP183730
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Human Lymphotoxin-alpha/TNF-beta ELISA Kit (Colorimetric) measures human Lymphotoxin-alpha/TNF-beta in cell culture supernatant, serum, plasma.

Specifications

Assay Range 15.6 - 1,000pg/mLmL
Assay Sensitivity 4.6pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 4049
Gene Symbol LTA
Quantity 96 Tests
Research Discipline Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym LT, LT-alpha, lymphotoxin alpha (TNF superfamily, member 1), TNFBlymphotoxin-alpha, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1
Target Lymphotoxin-alpha/TNF-beta
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.