Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lymphotoxin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB1370SP
This item is not returnable.
View return policy
Description
Lymphotoxin Monoclonal specifically detects Lymphotoxin in Human samples. It is validated for Neutralization.Specifications
Lymphotoxin | |
Monoclonal | |
LYOPH | |
Neutralization 0.15-0.9 ug/mL | |
LT, Lymphotoxin alpha, tnfb, TNFSF1, TNLG1E | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4049 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Neutralization | |
135125 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
LTA | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Lymphotoxin alpha/beta | |
25 μg | |
Primary | |
Recognizes human Lymphotoxin alpha chains in alpha 2 beta 1 or alpha 1 beta 2 trimers and also human TNF-beta . In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) APRIL, rhBAFF, rhEDA-A2, rhFas Ligand, rhLIGHT, rhGITR Ligand, rhOX40 Ligand, recombinant mouse RANKL, rhTNF-alpha , rhTRAIL, rhTRANCE, rhTWEAK, or rhVEGI is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction