Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Lymphotoxin beta R/TNFRSF3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6291100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6291100 100 μg
MAB6291SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6291100 Supplier R&D Systems Supplier No. MAB6291100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Agonist Activity.

Specifications

Antigen Lymphotoxin beta R/TNFRSF3
Classification Monoclonal
Clone 71315
Conjugate Unconjugated
Dilution Agonist Activity 1.5-9 ng/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P36941
Gene Alias CD18, D12S370, ltbetar, LT-BETA-R, LTBR, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, LymphotoxinbR, TNF RIII, TNF Rrp, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Lymphotoxin beta R/TNFRSF3 Gln31-Met227 Accession # P36941
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Research Discipline Agonist Activity
Primary or Secondary Primary
Gene ID (Entrez) 4055
Test Specificity Detects human Lymphotoxin beta R/TNFRSF3 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.